Assay Details
Target Gene Details
Entrez Gene ID: | 92675 |
Gene Name: | D-tyrosyl-tRNA deacylase 1 |
Gene Aliases: |
C20orf88, DUE-B, DUEB, HARS2, pqn-68 |
Location: |
Chr.20:18587893-18763917 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTD1 | NM_080820.5 | NP_543010.3 | ||
| XM_005260872.4 | XP_005260929.1 | |||
| AF332356.1 | AAL57046.1 | |||
| AK291440.1 | ||||
| BC045167.2 | AAH45167.1 | |||
| BC100923.1 | AAI00924.1 | |||
| BC100924.2 | AAI00925.1 | |||
| BC100925.1 | AAI00926.1 | |||
| BC100926.1 | AAI00927.1 |
Target Gene Details
Entrez Gene ID: | 101929526 |
Gene Name: | uncharacterized LOC101929526 |
Gene Aliases: |
- |
Location: |
Chr.20:18674395-18698708 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101929526 | NR_109955.1 | 1 | 113 |
More Information
Set Membership: |
|

Genomic Map