Assay Details
Target Gene Details
Entrez Gene ID: | 101927631 |
Gene Name: | ADNP antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:50930984-50945134 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADNP-AS1 | NR_110007.1 | 4 | 826 | |
| NR_110008.1 | 3 | 725 | ||
| NR_110009.1 | 3 | 722 | ||
| BX105402.1 | 1 | 451 |
Target Gene Details
Entrez Gene ID: | 8813 |
Gene Name: | dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit |
Gene Aliases: |
CDGIE, MPDS |
Location: |
Chr.20:50934855-50958564 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPM1 | NM_001317034.1 | NP_001303963.1 | ||
| NM_001317035.1 | NP_001303964.1 | |||
| NM_001317036.1 | NP_001303965.1 | |||
| NM_003859.2 | NP_003850.1 | |||
| NR_133648.1 | ||||
| XM_011529093.2 | XP_011527395.1 | |||
| AF007875.1 | AAC98797.1 | |||
| AK289569.1 | ||||
| BC007073.1 | AAH07073.1 | |||
| BC008466.1 | AAH08466.1 | |||
| BC016322.1 | AAH16322.1 | |||
| BM922866.1 | ||||
| BQ441542.1 | ||||
| CR456926.1 | CAG33207.1 | |||
| D86198.1 | BAA25646.1 | |||
| DR156925.1 | ||||
| KU178444.1 | ||||
| KU178445.1 | ||||
| KU178446.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834002 | Chr.20:50810235 - 50986764 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map