Assay Details
Target Gene Details
Entrez Gene ID: | 11142 |
Gene Name: | protein kinase (cAMP-dependent, catalytic) inhibitor gamma |
Gene Aliases: |
PKI-gamma |
Location: |
Chr.20:44531781-44619037 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKIG | NM_001281444.1 | NP_001268373.1 | ||
| NM_007066.4 | NP_008997.1 | |||
| NM_181804.2 | NP_861520.1 | |||
| NM_181805.2 | NP_861521.1 | |||
| XM_017027615.1 | XP_016883104.1 | |||
| AF182032.1 | AAD55445.1 | |||
| AK312048.1 | ||||
| BC104256.1 | AAI04257.1 | |||
| BC104257.1 | AAI04258.1 | |||
| BF569812.1 | ||||
| BG717090.1 | ||||
| BI461377.1 | ||||
| BU161358.1 | ||||
| CD244921.1 | ||||
| CN354542.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1061719 | Chr.20:44415722 - 44602319 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map