Assay Details
Target Gene Details
Entrez Gene ID: | 128434 |
Gene Name: | V-set and transmembrane domain containing 2 like |
Gene Aliases: |
C20orf102, dJ1118M15.2 |
Location: |
Chr.20:37903097-37945350 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VSTM2L | NM_080607.2 | NP_542174.1 | ||
| XM_011528530.1 | XP_011526832.1 | |||
| AK056177.1 | BAB71113.1 | |||
| AL834410.1 | CAD39072.2 | |||
| BC033818.1 | AAH33818.1 | |||
| BE905791.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2633566 | Chr.20:37913769 - 37914815 on Build GRCh38 | Insertion |
|
| nsv1066778 | Chr.20:37849676 - 37921323 on Build GRCh38 | Gain |
|
| nsv1057334 | Chr.20:37793679 - 38139909 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map