Assay Details
Target Gene Details
Entrez Gene ID: | 55902 |
Gene Name: | acyl-CoA synthetase short-chain family member 2 |
Gene Aliases: |
ACAS2, ACECS, ACS, ACSA, dJ1161H23.1 |
Location: |
Chr.20:34874942-34927966 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSS2 | NM_001076552.2 | NP_001070020.2 | ||
| NM_001242393.1 | NP_001229322.1 | |||
| NM_018677.3 | NP_061147.1 | |||
| XM_005260455.1 | XP_005260512.1 | |||
| XM_005260456.1 | XP_005260513.1 | |||
| XM_006723826.1 | XP_006723889.1 | |||
| XM_011528905.1 | XP_011527207.1 | |||
| XM_011528906.1 | XP_011527208.1 | |||
| XM_011528907.1 | XP_011527209.1 | |||
| XM_011528908.1 | XP_011527210.1 | |||
| XM_011528909.2 | XP_011527211.1 | |||
| XM_011528911.1 | XP_011527213.1 | |||
| XM_011528912.2 | XP_011527214.1 | |||
| AF263614.1 | AAF75064.1 | |||
| AK000162.1 | ||||
| AK000188.1 | ||||
| AK025990.1 | ||||
| AK092281.1 | BAC03849.1 | |||
| AK098026.1 | ||||
| AK293634.1 | ||||
| BC012172.1 | AAH12172.1 | |||
| BC073846.1 | AAH73846.1 | |||
| DA411343.1 | ||||
| DA626707.1 | ||||
| DA628275.1 | ||||
| DA630609.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055447 | Chr.20:34737641 - 35020344 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map