Assay Details
Target Gene Details
Entrez Gene ID: | 92675 |
Gene Name: | D-tyrosyl-tRNA deacylase 1 |
Gene Aliases: |
C20orf88, DUE-B, DUEB, HARS2, pqn-68 |
Location: |
Chr.20:18587893-18763917 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTD1 | NM_001318043.1 | NP_001304972.1 | ||
| NM_080820.5 | NP_543010.3 | |||
| XM_005260872.4 | XP_005260929.1 | |||
| AF332356.1 | AAL57046.1 | |||
| AK074304.1 | BAB85044.1 | |||
| AK291440.1 | ||||
| BC045167.2 | AAH45167.1 | |||
| BC100923.1 | AAI00924.1 | |||
| BC100924.2 | AAI00925.1 | |||
| BC100925.1 | AAI00926.1 | |||
| BC100926.1 | AAI00927.1 | |||
| BI159760.1 | ||||
| BI465352.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4283n100 | Chr.20:18364565 - 18647377 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map