Assay Details
Target Gene Details
Entrez Gene ID: | 55902 |
Gene Name: | acyl-CoA synthetase short-chain family member 2 |
Gene Aliases: |
ACAS2, ACECS, ACS, ACSA, dJ1161H23.1 |
Location: |
Chr.20:34874942-34927966 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055447 | Chr.20:34737641 - 35020344 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map