Assay Details
Target Gene Details
Entrez Gene ID: | 200232 |
Gene Name: | family with sequence similarity 209 member A |
Gene Aliases: |
C20orf106, dJ1153D9.3 |
Location: |
Chr.20:56524729-56533590 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM209A |
Target Gene Details
Entrez Gene ID: | 388799 |
Gene Name: | family with sequence similarity 209 member B |
Gene Aliases: |
C20orf107, dJ1153D9.4 |
Location: |
Chr.20:56527124-56555626 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM209B | XM_017027849.1 | XP_016883338.1 | ||
| BC105792.1 | AAI05793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2722576 | Chr.20:56526267 - 56536764 on Build GRCh38 | Deletion |
|
| nsv525425 | Chr.20:56403979 - 56897294 on Build GRCh38 | Gain |
|
| nsv834015 | Chr.20:56518712 - 56656502 on Build GRCh38 | Loss |
|
| nsv1056574 | Chr.20:56495687 - 56761211 on Build GRCh38 | Gain |
|

Genomic Map