Assay Details
Target Gene Details
Entrez Gene ID: | 3872 |
Gene Name: | keratin 17 |
Gene Aliases: |
39.1, CK-17, K17, PC, PC2, PCHC1 |
Location: |
Chr.17:41619440-41624630 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT17 | NM_000422.2 | NP_000413.1 | ||
| AK095342.1 | BAC04534.1 | |||
| AK304373.1 | ||||
| BC000159.2 | AAH00159.2 | |||
| BC011901.2 | AAH11901.1 | |||
| BC056421.1 | AAH56421.1 | |||
| BC072018.1 | AAH72018.1 | |||
| BC072019.1 | AAH72019.1 | |||
| BP226061.1 | ||||
| BX647923.1 | ||||
| X62571.1 | CAA44451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34903 | Chr.17:41489914 - 41632423 on Build GRCh38 | Gain |
|
| nsv962326 | Chr.17:41608774 - 41632235 on Build GRCh38 | Duplication |
|
| esv3640591 | Chr.17:41605673 - 41624921 on Build GRCh38 | Gain |
|
| nsv575040 | Chr.17:41610982 - 41624576 on Build GRCh38 | Gain |
|
| nsv827992 | Chr.17:41488451 - 41630341 on Build GRCh38 | Gain |
|
| esv2758690 | Chr.17:41145123 - 41740591 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map