Assay Details
Target Gene Details
Entrez Gene ID: | 29110 |
Gene Name: | TANK binding kinase 1 |
Gene Aliases: |
FTDALS4, NAK, T2K |
Location: |
Chr.12:64452060-64502119 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBK1 | NM_013254.3 | NP_037386.1 | ||
| XM_005268809.1 | XP_005268866.1 | |||
| AF191838.1 | AAF05989.1 | |||
| AK291039.1 | ||||
| BC034950.2 | AAH34950.1 | |||
| DA788357.1 |
More Information
Set Membership: |
|

Genomic Map