Assay Details
Target Gene Details
Entrez Gene ID: | 643382 |
Gene Name: | transmembrane protein 253 |
Gene Aliases: |
C14orf176, C14orf95, NCRNA00220 |
Location: |
Chr.14:21098937-21103724 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM253 | NM_001146683.1 | 8 | 1406 | NP_001140155.1 |
| XM_006720233.3 | 7 | 1981 | XP_006720296.1 | |
| XM_011537078.2 | 8 | 1283 | XP_011535380.1 | |
| XM_011537079.1 | 7 | 1125 | XP_011535381.1 | |
| XM_011537081.2 | 6 | 1872 | XP_011535383.1 | |
| XM_011537082.2 | 7 | 1290 | XP_011535384.1 | |
| XM_011537083.2 | 6 | 1873 | XP_011535385.1 | |
| XM_011537084.2 | 6 | 1845 | XP_011535386.1 | |
| XM_011537085.2 | 5 | 1765 | XP_011535387.1 | |
| AA284845.1 | 1 | 120 |
Target Gene Details
Entrez Gene ID: | 51222 |
Gene Name: | zinc finger protein 219 |
Gene Aliases: |
ZFP219 |
Location: |
Chr.14:21090046-21104704 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF219 | NM_001102454.1 | NP_001095924.1 | ||
| DA013397.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv123n21 | Chr.14:21072704 - 21106531 on Build GRCh38 | Loss |
|
| nsv826882 | Chr.14:21042862 - 21136042 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map