Assay Details
Target Gene Details
Entrez Gene ID: | 10425 |
Gene Name: | ariadne RBR E3 ubiquitin protein ligase 2 |
Gene Aliases: |
ARI2, TRIAD1 |
Location: |
Chr.3:48918820-48985541 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Gene Details
Entrez Gene ID: | 646450 |
Gene Name: | ariadne homolog 2 opposite strand |
Gene Aliases: |
C3orf71 |
Location: |
Chr.3:48917788-48919385 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARIH2OS | NM_001123040.1 | 1 | 346 | NP_001116512.1 |
| AK097724.1 | 1 | 346 | BAC05151.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893694 | Chr.3:48792101 - 48921349 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map