Assay Details
Target Gene Details
Entrez Gene ID: | 84303 |
Gene Name: | coiled-coil-helix-coiled-coil-helix domain containing 6 |
Gene Aliases: |
CHCM1, Mic25, PPP1R23 |
Location: |
Chr.3:126704220-126960420 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHCHD6 | NM_001320610.1 | NP_001307539.1 | ||
| NM_032343.2 | NP_115719.1 | |||
| XM_011513234.1 | XP_011511536.1 | |||
| XM_011513235.2 | XP_011511537.1 | |||
| XM_017007320.1 | XP_016862809.1 | |||
| XM_017007321.1 | XP_016862810.1 | |||
| XM_017007322.1 | XP_016862811.1 | |||
| AK310910.1 | ||||
| AK311056.1 | ||||
| BC006123.1 | AAH06123.1 | |||
| DN992600.1 | ||||
| JF264889.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33263 | Chr.3:126707697 - 126708380 on Build GRCh38 | Gain |
|
| nsv829718 | Chr.3:126629911 - 126809810 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map