Assay Details
Target Gene Details
Entrez Gene ID: | 79903 |
Gene Name: | N(alpha)-acetyltransferase 60, NatF catalytic subunit |
Gene Aliases: |
HAT4, NAT15 |
Location: |
Chr.16:3443611-3486963 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA60 | NM_001083601.2 | NP_001077070.1 | ||
| NM_001317093.1 | NP_001304022.1 | |||
| AK297219.1 | ||||
| AL833594.1 | ||||
| DA786716.1 | ||||
| DC401343.1 |
More Information
Set Membership: |
|

Genomic Map