Assay Details
Target Gene Details
Entrez Gene ID: | 113829 |
Gene Name: | solute carrier family 35 member A4 |
Gene Aliases: |
- |
Location: |
Chr.5:140564565-140569104 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35A4 | NM_080670.3 | NP_542401.1 | ||
| AF445025.1 | 1 | 752 | AAL38586.1 | |
| AK122601.1 | BAC85488.1 | |||
| AK307282.1 | ||||
| BC009906.2 | AAH09906.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv462462 | Chr.5:140556654 - 140626201 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map