Assay Details
Target Gene Details
Entrez Gene ID: | 3156 |
Gene Name: | 3-hydroxy-3-methylglutaryl-CoA reductase |
Gene Aliases: |
LDLCQ3 |
Location: |
Chr.5:75336334-75362101 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMGCR | NM_000859.2 | NP_000850.1 | ||
| NM_001130996.1 | NP_001124468.1 | |||
| XM_011543357.1 | XP_011541659.1 | |||
| XM_011543359.1 | XP_011541661.1 | |||
| AK296499.1 | ||||
| M62627.1 | 1 | 184 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575856 | Chr.5:75260838 - 75400813 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map