Assay Details
Target Gene Details
Entrez Gene ID: | 57128 |
Gene Name: | LYR motif containing 4 |
Gene Aliases: |
C6orf149, CGI-203, COXPD19, ISD11 |
Location: |
Chr.6:5031029-5260950 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYRM4 | NM_001318783.1 | NP_001305712.1 | ||
| NR_134856.1 | ||||
| XM_017011083.1 | XP_016866572.1 | |||
| XM_017011084.1 | XP_016866573.1 | |||
| DA114665.1 | ||||
| DA222333.1 |
Target Gene Details
Entrez Gene ID: | 100129461 |
Gene Name: | LYRM4 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.6:5003816-5240496 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYRM4-AS1 | NR_126015.1 | |||
| AK094934.1 |
Target Gene Details
Entrez Gene ID: | 648791 |
Gene Name: | protein phosphatase 1 regulatory subunit 3G |
Gene Aliases: |
- |
Location: |
Chr.6:5085486-5087221 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R3G | NM_001145115.2 | 1 | 511 | NP_001138587.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600871 | Chr.6:4904146 - 5171775 on Build GRCh38 | Gain |
|
| nsv600872 | Chr.6:4975079 - 5341957 on Build GRCh38 | Gain |
|
| nsv1017380 | Chr.6:5007636 - 5152317 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map