Assay Details
Target Gene Details
Entrez Gene ID: | 123745 |
Gene Name: | phospholipase A2 group IVE |
Gene Aliases: |
- |
Location: |
Chr.15:41981582-42050703 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G4E | NM_001206670.1 | 20 | 2455 | NP_001193599.1 |
| XM_011521237.2 | 20 | 2365 | XP_011519539.1 | |
| XM_011521238.1 | 11 | 2173 | XP_011519540.1 | |
| XM_011521239.1 | 9 | 1301 | XP_011519541.1 | |
| AK127558.1 | 10 | 1744 | BAC87034.1 | |
| BC101584.1 | 9 | 1360 | ||
| BC101612.1 | 9 | 1360 | ||
| BX648482.1 | 10 | 1654 |
Target Gene Details
Entrez Gene ID: | 101928388 |
Gene Name: | PLA2G4E antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.15:41972763-41999094 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G4E-AS1 | NR_120334.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain |
|
| nsv1038933 | Chr.15:41846258 - 42016889 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map