Assay Details
Target Gene Details
Entrez Gene ID: | 117581 |
Gene Name: | twist family bHLH transcription factor 2 |
Gene Aliases: |
AMS, BBRSAY, DERMO1, FFDD3, SETLSS, bHLHa39 |
Location: |
Chr.2:238848032-238910548 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TWIST2 | NM_001271893.3 | 2 | 1190 | NP_001258822.1 |
| BC017907.1 | 2 | 1137 | AAH17907.1 | |
| BC103755.1 | 2 | 1146 | AAI03756.1 | |
| DB359681.1 | 1 | 220 |
More Information
Set Membership: |
|

Genomic Map