Assay Details
Target Gene Details
Entrez Gene ID: | 101927139 |
Gene Name: | uncharacterized LOC101927139 |
Gene Aliases: |
- |
Location: |
Chr.1:66042500-66050718 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927139 | NR_123718.1 | |||
| BX109047.1 |
Target Gene Details
Entrez Gene ID: | 5142 |
Gene Name: | phosphodiesterase 4B |
Gene Aliases: |
DPDE4, PDEIVB |
Location: |
Chr.1:65792510-66374579 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE4B | NM_001037340.2 | NP_001032417.1 | ||
| NM_001037341.1 | NP_001032418.1 | |||
| NM_001297440.1 | NP_001284369.1 | |||
| NM_001297441.1 | NP_001284370.1 | |||
| NM_002600.3 | NP_002591.2 | |||
| XM_011541566.1 | XP_011539868.1 | |||
| XM_017001446.1 | XP_016856935.1 | |||
| AB209081.1 | BAD92318.1 | |||
| AK289969.1 | ||||
| AK290006.1 | ||||
| BC101480.1 | AAI01481.1 | |||
| BC105040.1 | AAI05041.1 | |||
| BF513098.1 | ||||
| CR749667.1 | CAH18458.2 | |||
| DA103627.1 | ||||
| DA126733.1 | ||||
| DA332822.1 | ||||
| KJ534916.1 | ||||
| KJ534917.1 | ||||
| KJ535082.1 | ||||
| L20966.1 | AAA03589.1 | |||
| U85048.1 | AAB96381.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3365990 | Chr.1:66045712 - 66045750 on Build GRCh38 | Insertion |
|
| nsv998127 | Chr.1:66038974 - 66069548 on Build GRCh38 | Loss |
|
| esv3306072 | Chr.1:66045700 - 66045701 on Build GRCh38 | Mobile Element Insertion |
|
More Information
Set Membership: |
|

Genomic Map