Assay Details
Target Gene Details
Entrez Gene ID: | 100128675 |
Gene Name: | HPN antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:35059059-35106304 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HPN-AS1 | NR_024561.1 | 4 | 957 | |
| NR_024562.1 | ||||
| AK124779.1 | 4 | 957 | BAC85944.1 | |
| AK295317.1 |

Genomic Map