Assay Details
Target Gene Details
Entrez Gene ID: | 200232 |
Gene Name: | family with sequence similarity 209 member A |
Gene Aliases: |
C20orf106, dJ1153D9.3 |
Location: |
Chr.20:56524729-56533590 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM209A | NM_001012971.3 | NP_001012989.2 | ||
| BC029411.1 | ||||
| BC107704.1 | AAI07705.1 | |||
| BU738481.1 | ||||
| BX335293.2 |
Target Gene Details
Entrez Gene ID: | 140687 |
Gene Name: | glucosaminyl (N-acetyl) transferase family member 7 |
Gene Aliases: |
C20orf105, dJ1153D9.2, gcnt |
Location: |
Chr.20:56491492-56525925 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCNT7 | NM_080615.1 | NP_542182.1 | ||
| AK131203.1 | BAD18395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525425 | Chr.20:56403979 - 56897294 on Build GRCh38 | Gain |
|
| nsv834015 | Chr.20:56518712 - 56656502 on Build GRCh38 | Loss |
|
| nsv1056574 | Chr.20:56495687 - 56761211 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map