Assay Details
Target Gene Details
Entrez Gene ID: | 391257 |
Gene Name: | SUMO1 pseudogene 1 |
Gene Aliases: |
PIC1L, UBL2, UBL6 |
Location: |
Chr.20:53874498-53875709 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUMO1P1 | NR_002189.3 | 1 | 147 | |
| BC065723.1 | 1 | 150 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1060372 | Chr.20:53792469 - 53885561 on Build GRCh38 | Loss |
|
| esv34060 | Chr.20:53635324 - 54085647 on Build GRCh38 | Loss |
|
| nsv586224 | Chr.20:53858311 - 54662951 on Build GRCh38 | Gain |
|
| dgv66n68 | Chr.20:53752121 - 53974256 on Build GRCh38 | Loss |
|
| nsv586222 | Chr.20:53857140 - 53889841 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116095814] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map