Assay Details
Target Gene Details
Entrez Gene ID: | 8447 |
Gene Name: | double C2 domain beta |
Gene Aliases: |
DOC2BL |
Location: |
Chr.17:146748-181630 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOC2B | NM_003585.4 | NP_003576.2 | ||
| XM_011535363.2 | XP_011533665.1 | |||
| XM_017025227.1 | XP_016880716.1 | |||
| AI478508.1 | ||||
| D70830.1 | BAA11107.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5346n54 | Chr.17:168352 - 181795 on Build GRCh38 | Loss |
|
| nsv573862 | Chr.17:153794 - 184412 on Build GRCh38 | Gain |
|
| nsv469538 | Chr.17:150208 - 266971 on Build GRCh38 | Loss |
|
| nsv573876 | Chr.17:170860 - 178833 on Build GRCh38 | Gain |
|
| esv3639614 | Chr.17:176972 - 217320 on Build GRCh38 | Gain |
|
| dgv3068n100 | Chr.17:150721 - 222292 on Build GRCh38 | Gain |
|
| dgv5347n54 | Chr.17:168571 - 184412 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map