Assay Details
Target Gene Details
Entrez Gene ID: | 6358 |
Gene Name: | C-C motif chemokine ligand 14 |
Gene Aliases: |
CC-1, CC-3, CKB1, HCC-1, HCC-1(1-74), HCC-1/HCC-3, HCC-3, MCIF, NCC-2, NCC2, SCYA14, SCYL2, SY14 |
Location: |
Chr.17:35983656-35986728 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL14 | NM_032962.4 | 4 | 396 | NP_116738.1 |
| NM_032963.3 | 3 | 348 | NP_116739.1 | |
| AA612866.1 | 1 | 160 | ||
| AK122606.1 | 2 | 1198 | ||
| BC038289.1 | 3 | 313 | AAH38289.1 | |
| BC045165.1 | 3 | 330 | AAH45165.1 | |
| BI496964.1 | ||||
| DA821276.1 | 3 | 350 | ||
| Z49270.1 | 3 | 323 | CAA89264.1 |
Target Gene Details
Entrez Gene ID: | 348249 |
Gene Name: | CCL15-CCL14 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.17:35983656-36002038 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCL15-CCL14 | NR_027921.3 | 8 | 1306 | |
| NR_027922.3 | 7 | 1258 | ||
| Z70292.1 | 7 | 767 | ||
| Z70293.1 | 8 | 815 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059795 | Chr.17:35828724 - 36157706 on Build GRCh38 | Gain |
|
| nsv833427 | Chr.17:35958281 - 36115227 on Build GRCh38 | Loss |
|
| nsv833424 | Chr.17:35911883 - 36092128 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map