Assay Details
Target Gene Details
Entrez Gene ID: | 58526 |
Gene Name: | MID1 interacting protein 1 |
Gene Aliases: |
G12-like, MIG12, S14R, STRAIT11499, THRSPL |
Location: |
Chr.X:38801432-38806532 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MID1IP1 | NM_001098790.1 | NP_001092260.1 | ||
| NM_001098791.1 | NP_001092261.1 | |||
| NM_021242.5 | 2 | 1208 | NP_067065.1 | |
| BC019332.1 | AAH19332.1 | |||
| CN417714.1 | 1 | 610 | ||
| DA138791.1 |
Target Gene Details
Entrez Gene ID: | 100874211 |
Gene Name: | MID1IP1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.X:38801248-38803883 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MID1IP1-AS1 | NR_046706.1 | |||
| DY655415.1 |
More Information
Set Membership: |
|

Genomic Map