Assay Details
Target Gene Details
Entrez Gene ID: | 2934 |
Gene Name: | gelsolin |
Gene Aliases: |
ADF, AGEL |
Location: |
Chr.9:121201082-121332844 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Target Gene Details
Entrez Gene ID: | 57000 |
Gene Name: | GSN antisense RNA 1 |
Gene Aliases: |
C9orf31, MOST2 |
Location: |
Chr.9:121280768-121285530 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSN-AS1 | NR_103560.1 | 1 | 1073 | |
| AF220263.1 | 1 | 1073 | AAF89164.1 |
More Information
Set Membership: |
|

Genomic Map