Assay Details
Target Gene Details
Entrez Gene ID: | 1956 |
Gene Name: | epidermal growth factor receptor |
Gene Aliases: |
ERBB, ERBB1, HER1, NISBD2, PIG61, mENA |
Location: |
Chr.7:55019032-55207338 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EGFR | NM_005228.3 | NP_005219.2 | ||
| AB209442.1 | BAD92679.1 | |||
| AK225422.1 | ||||
| AK290352.1 | ||||
| AK294750.1 | ||||
| AU137334.1 | ||||
| AY550966.1 | AAT52212.1 | |||
| BC094761.1 | AAH94761.1 | |||
| DQ088980.1 | ||||
| GU255993.1 | ||||
| HM437234.1 | ||||
| HQ912715.1 | ||||
| K03193.1 | AAA52371.1 | |||
| X00588.1 | CAA25240.1 | |||
| X00663.1 | CAA25282.1 |
Target Gene Details
Entrez Gene ID: | 100507500 |
Gene Name: | EGFR antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:55179750-55188949 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EGFR-AS1 | NR_047551.1 | 2 | 283 | |
| AA004947.1 | 2 | 288 | ||
| AK123474.1 | 2 | 268 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734425 | Chr.7:53868335 - 55917254 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map