Assay Details
Target Gene Details
Entrez Gene ID: | 93664 |
Gene Name: | calcium dependent secretion activator 2 |
Gene Aliases: |
CAPS2 |
Location: |
Chr.7:122318424-122886759 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Target Gene Details
Entrez Gene ID: | 378925 |
Gene Name: | ring finger protein 148 |
Gene Aliases: |
- |
Location: |
Chr.7:122701666-122702967 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF148 | NM_198085.1 | 1 | 839 | NP_932351.1 |
| AK058116.1 | ||||
| AK098654.1 | 1 | 797 | BAC05367.1 | |
| BC029264.1 | 1 | 839 | AAH29264.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3891198 | Chr.7:121885430 - 123886905 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map