Assay Details
Target Gene Details
Entrez Gene ID: | 285965 |
Gene Name: | EPHA1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.7:143407813-143523449 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA1-AS1 | NR_033897.1 | |||
| AL833583.1 |
Target Gene Details
Entrez Gene ID: | 259287 |
Gene Name: | taste 2 receptor member 41 |
Gene Aliases: |
T2R41, T2R59 |
Location: |
Chr.7:143477873-143478796 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAS2R41 | NM_176883.2 | 1 | 248 | NP_795364.2 |
| BC101157.3 | 1 | 248 | AAI01158.1 | |
| BC101158.2 | 1 | 248 | AAI01159.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428188 | Chr.7:143450682 - 144500577 on Build GRCh38 | Gain+Loss |
|
| esv3891218 | Chr.7:143457550 - 143479566 on Build GRCh38 | Loss |
|
| nsv608682 | Chr.7:143444382 - 144177097 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map