Assay Details
Target Gene Details
Entrez Gene ID: | 101154753 |
Gene Name: | promoter of CDKN1A antisense DNA damage activated RNA |
Gene Aliases: |
PANDA |
Location: |
Chr.6:36673621-36675126 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PANDAR | NR_109836.1 | 1 | 1215 | |
| JF803844.1 | 1 | 1215 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26139 | Chr.6:36672399 - 36674405 on Build GRCh38 | Loss |
|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|

Genomic Map