Assay Details
Target Gene Details
Entrez Gene ID: | 51527 |
Gene Name: | GSK3B interacting protein |
Gene Aliases: |
C14orf129, HSPC210 |
Location: |
Chr.14:96363452-96387290 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GSKIP | NM_001271904.1 | NP_001258833.1 | ||
| NM_001271905.1 | NP_001258834.1 | |||
| NM_001271906.1 | NP_001258835.1 | |||
| NM_016472.4 | NP_057556.2 | |||
| XM_017021365.1 | XP_016876854.1 | |||
| AF151044.1 | AAF36130.1 | |||
| AK000796.1 | BAA91380.1 | |||
| AK094654.1 | ||||
| AK314423.1 | ||||
| DA488110.1 | ||||
| HY019704.1 | ||||
| HY051680.1 | ||||
| HY108584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2660133 | Chr.14:96364806 - 96366404 on Build GRCh38 | Deletion |
|
| esv3552010 | Chr.14:96364893 - 96366347 on Build GRCh38 | Deletion |
|
| esv28071 | Chr.14:96365009 - 96366314 on Build GRCh38 | Loss |
|
| esv3635422 | Chr.14:96364866 - 96366338 on Build GRCh38 | Loss |
|

Genomic Map