Assay Details
Target Gene Details
Entrez Gene ID: | 4008 |
Gene Name: | LIM domain 7 |
Gene Aliases: |
FBX20, FBXO20, LOMP |
Location: |
Chr.13:75620434-75859870 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMO7 | NM_001306080.1 | NP_001293009.1 | ||
| NM_005358.5 | NP_005349.3 | |||
| AF330045.1 | AAL37480.1 | |||
| AK092052.1 | ||||
| DA313248.1 | ||||
| FJ711162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751152 | Chr.13:75570063 - 75714282 on Build GRCh38 | Loss |
|
| nsv521444 | Chr.13:74139302 - 75679218 on Build GRCh38 | Gain |
|
| dgv3260n54 | Chr.13:75636175 - 75636921 on Build GRCh38 | Loss |
|
| dgv3257n54 | Chr.13:75635643 - 75636856 on Build GRCh38 | Gain |
|
| nsv562359 | Chr.13:75636005 - 75636718 on Build GRCh38 | Gain |
|
| esv22314 | Chr.13:75635563 - 75636982 on Build GRCh38 | Loss |
|
| dgv3259n54 | Chr.13:75635745 - 75636718 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map