Assay Details
Target Gene Details
Entrez Gene ID: | 140687 |
Gene Name: | glucosaminyl (N-acetyl) transferase family member 7 |
Gene Aliases: |
C20orf105, dJ1153D9.2, gcnt |
Location: |
Chr.20:56491492-56525925 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCNT7 | NM_080615.1 | NP_542182.1 | ||
| AK131203.1 | BAD18395.1 |
Target Gene Details
Entrez Gene ID: | 51507 |
Gene Name: | replication termination factor 2 domain containing 1 |
Gene Aliases: |
C20orf43, CDAO5, HSPC164, SHUJUN-3 |
Location: |
Chr.20:56468585-56518886 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTFDC1 | NM_001283035.1 | NP_001269964.1 | ||
| NM_001283036.1 | NP_001269965.1 | |||
| NM_001283037.1 | NP_001269966.1 | |||
| NM_016407.4 | NP_057491.2 | |||
| XM_017027872.1 | XP_016883361.1 | |||
| AF117231.1 | AAF17212.1 | |||
| AF161513.1 | AAF29128.1 | |||
| AF161518.1 | AAF29133.1 | |||
| AF212244.1 | AAK14929.1 | |||
| AK000481.1 | BAA91193.1 | |||
| AK000586.1 | ||||
| AK026118.1 | ||||
| AK225135.1 | ||||
| AK293662.1 | ||||
| AK302029.1 | ||||
| AK314563.1 | ||||
| BC003359.1 | AAH03359.1 | |||
| BG468509.1 | ||||
| BU537939.1 | ||||
| CR983692.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525425 | Chr.20:56403979 - 56897294 on Build GRCh38 | Gain |
|
| esv29810 | Chr.20:56495233 - 56495910 on Build GRCh38 | Gain |
|
| nsv1056574 | Chr.20:56495687 - 56761211 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map