Assay Details
Target Gene Details
Entrez Gene ID: | 490 |
Gene Name: | ATPase plasma membrane Ca2+ transporting 1 |
Gene Aliases: |
PMCA1, PMCA1kb |
Location: |
Chr.12:89588049-89709353 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP2B1 | XM_011538407.2 | XP_011536709.1 |
Target Gene Details
Entrez Gene ID: | 338758 |
Gene Name: | long intergenic non-protein coding RNA 936 |
Gene Aliases: |
- |
Location: |
Chr.12:89708955-89711952 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LINC00936 | NR_028138.1 | 1 | 245 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2775n54 | Chr.12:89708524 - 89709462 on Build GRCh38 | Loss |
|
| nsv559720 | Chr.12:89709029 - 89709254 on Build GRCh38 | Loss |
|
| esv28180 | Chr.12:89708163 - 89709628 on Build GRCh38 | Loss |
|
| dgv2773n54 | Chr.12:89708316 - 89709574 on Build GRCh38 | Gain |
|
| nsv559705 | Chr.12:89702019 - 89713058 on Build GRCh38 | Gain |
|
| dgv2776n54 | Chr.12:89708819 - 89709574 on Build GRCh38 | Gain+Loss |
|
| nsv559709 | Chr.12:89708471 - 89713058 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map