Assay Details
Target Gene Details
Entrez Gene ID: | 8835 |
Gene Name: | suppressor of cytokine signaling 2 |
Gene Aliases: |
CIS2, Cish2, SOCS-2, SSI-2, SSI2, STATI2 |
Location: |
Chr.12:93569822-93626236 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS2 | NM_001270467.1 | NP_001257396.1 | ||
| NM_003877.4 | NP_003868.1 | |||
| XM_017020147.1 | XP_016875636.1 | |||
| XM_017020152.1 | XP_016875641.1 | |||
| AK313165.1 | ||||
| AL522912.3 | ||||
| DA709082.1 | 1 | 242 |
Target Gene Details
Entrez Gene ID: | 144481 |
Gene Name: | SOCS2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:93565628-93571398 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS2-AS1 | NR_038263.1 | |||
| AK054607.1 | ||||
| AK098304.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25615 | Chr.12:93570606 - 93572157 on Build GRCh38 | Loss |
|
| nsv559778 | Chr.12:93570630 - 93571360 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map