Assay Details
Target Gene Details
Entrez Gene ID: | 26039 |
Gene Name: | SS18L1, nBAF chromatin remodeling complex subunit |
Gene Aliases: |
CREST, LP2261 |
Location: |
Chr.20:62143720-62182510 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SS18L1 | NM_001301778.1 | NP_001288707.1 | ||
| NM_198935.2 | NP_945173.1 | |||
| NR_125980.1 | ||||
| NR_125981.1 | ||||
| XM_005260389.3 | XP_005260446.1 | |||
| XM_005260391.1 | XP_005260448.1 | |||
| XM_017027784.1 | 1 | 178 | XP_016883273.1 | |
| AB014593.1 | ||||
| AK125656.1 | ||||
| AK291485.1 | ||||
| AK299881.1 | ||||
| AK308765.1 | ||||
| BX640848.1 | CAE45918.1 | |||
| DC375156.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509782 | Chr.20:62125636 - 62180328 on Build GRCh38 | Insertion |
|
| dgv7648n54 | Chr.20:62143364 - 62144394 on Build GRCh38 | Gain+Loss |
|
| nsv586476 | Chr.20:62140816 - 62144394 on Build GRCh38 | Gain |
|
| esv22512 | Chr.20:62143349 - 62144455 on Build GRCh38 | Loss |
|
| dgv7649n54 | Chr.20:62143468 - 62144394 on Build GRCh38 | Gain |
|
| nsv3440 | Chr.20:62137705 - 62169472 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map