Assay Details
Target Gene Details
Entrez Gene ID: | 2900 |
Gene Name: | glutamate ionotropic receptor kainate type subunit 4 |
Gene Aliases: |
EAA1, GRIK, GluK4, KA1 |
Location: |
Chr.11:120511746-120988805 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRIK4 | NM_014619.4 | NP_055434.2 | ||
| AK292726.1 | ||||
| DA246271.1 | ||||
| DA740816.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1150134 | Chr.11:120516992 - 120517668 on Build GRCh38 | Deletion |
|
| esv22400 | Chr.11:120517092 - 120517672 on Build GRCh38 | Loss |
|
| esv3627905 | Chr.11:120516925 - 120517652 on Build GRCh38 | Loss |
|
| esv3548428 | Chr.11:120516826 - 120517746 on Build GRCh38 | Deletion |
|
| dgv1289n100 | Chr.11:120506099 - 120595091 on Build GRCh38 | Gain |
|
| nsv556474 | Chr.11:120517119 - 120520764 on Build GRCh38 | Loss |
|
| nsv556472 | Chr.11:120516490 - 120517627 on Build GRCh38 | Loss |
|
| esv2666173 | Chr.11:120516931 - 120517657 on Build GRCh38 | Deletion |
|
| esv2745146 | Chr.11:120516941 - 120517651 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map