Assay Details
Target Gene Details
Entrez Gene ID: | 9989 |
Gene Name: | protein phosphatase 4 regulatory subunit 1 |
Gene Aliases: |
MEG1, PP4(Rmeg), PP4R1 |
Location: |
Chr.18:9546791-9615274 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP4R1 | XM_011525775.2 | XP_011524077.1 | ||
| XM_011525776.1 | XP_011524078.1 | |||
| XM_011525779.2 | XP_011524081.1 | |||
| XM_017026105.1 | XP_016881594.1 |
Target Gene Details
Entrez Gene ID: | 101927323 |
Gene Name: | PPP4R1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.18:9614807-9619363 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP4R1-AS1 | NR_110773.1 | 1 | 64 | |
| HY048010.1 | 1 | 65 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1072774 | Chr.18:9614401 - 9614902 on Build GRCh38 | Deletion |
|
| nsv1118782 | Chr.18:9614402 - 9615202 on Build GRCh38 | Deletion |
|
| nsv576448 | Chr.18:9558242 - 9828451 on Build GRCh38 | Gain |
|
| esv29650 | Chr.18:9614225 - 9614940 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map