Assay Details
Target Gene Details
Entrez Gene ID: | 84319 |
Gene Name: | cms1 ribosomal small subunit homolog (yeast) |
Gene Aliases: |
C3orf26 |
Location: |
Chr.3:99817834-100178632 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CMSS1 | NM_032359.3 | NP_115735.2 | ||
| AK124387.1 | ||||
| AK291392.1 | ||||
| AK312483.1 | ||||
| BC006475.1 | AAH06475.1 | |||
| BC006512.2 | AAH06512.1 |
Target Gene Details
Entrez Gene ID: | 11259 |
Gene Name: | filamin A interacting protein 1 like |
Gene Aliases: |
DOC-1, DOC1, GIP130, GIP90 |
Location: |
Chr.3:99833144-100114513 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FILIP1L | NM_001042459.2 | NP_001035924.1 | ||
| NM_182909.3 | NP_878913.2 | |||
| AF329092.1 | AAN16206.1 | |||
| AF514867.1 | AAQ08177.1 | |||
| AF514868.1 | AAQ08178.1 | |||
| AF514869.1 | AAQ08179.1 | |||
| AX776292.1 | CAE11726.1 | |||
| AX776294.1 | CAE11727.1 | |||
| AX776296.1 | CAE11728.1 | |||
| AX776298.1 | CAE11729.1 | |||
| AY642382.1 | AAV34207.1 | |||
| BC020941.1 | AAH20941.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3597161 | Chr.3:99909937 - 99910787 on Build GRCh38 | Loss |
|
| dgv8475n54 | Chr.3:99909978 - 99910723 on Build GRCh38 | Gain+Loss |
|
| esv998785 | Chr.3:99909957 - 99910760 on Build GRCh38 | Loss |
|
| esv3562278 | Chr.3:99909869 - 99910826 on Build GRCh38 | Deletion |
|
| esv2725688 | Chr.3:99909911 - 99910766 on Build GRCh38 | Deletion |
|
| nsv591147 | Chr.3:99910043 - 99910723 on Build GRCh38 | Loss |
|
| nsv954339 | Chr.3:99909942 - 99910786 on Build GRCh38 | Deletion |
|
| esv2945 | Chr.3:99909898 - 99910844 on Build GRCh38 | Loss |
|
| esv26006 | Chr.3:99909951 - 99910774 on Build GRCh38 | Loss |
|
| esv988265 | Chr.3:99909459 - 99911162 on Build GRCh38 | Deletion |
|
| esv987776 | Chr.3:99909938 - 99910786 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map