Assay Details
Target Gene Details
Entrez Gene ID: | 16183 |
Gene Name: | interleukin 2 |
Gene Aliases: |
Il-2 |
Location: |
Chr.3:37120713-37125954 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il2 | NM_008366.3 | NP_032392.1 | ||
| AF065914.1 | AAD25890.1 | |||
| AF065915.1 | AAD25891.1 | |||
| AF065916.1 | AAD25892.1 | |||
| AF538059.1 | AAN77291.1 | |||
| AF542383.1 | AAN77288.1 | |||
| AF542384.1 | AAN77289.1 | |||
| AF542385.1 | AAN77290.1 | |||
| AY147902.1 | AAN38301.1 | |||
| BC116845.1 | ||||
| BC116873.1 | ||||
| DQ836354.1 | ||||
| K02292.1 | AAA39289.1 | |||
| U41494.1 | AAB39206.1 | |||
| U41504.1 | AAB39208.1 | |||
| U41505.1 | AAB39209.1 | |||
| U41506.1 | AAB39210.1 | |||
| X01772.1 | CAA25909.1 | |||
| X66058.1 | CAA46854.1 | |||
| X73040.1 |
More Information
Set Membership: |
|

Genomic Map