Assay Details
Target Gene Details
Entrez Gene ID: | 224440 |
Gene Name: | SET domain containing 4 |
Gene Aliases: |
AY037804, C21orf18, ORF21 |
Location: |
Chr.16:93583457-93604833 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Setd4 | NM_145482.3 | 7 | 906 | NP_663457.2 |
| AK194589.1 | ||||
| AY037804.1 | 7 | 906 | AAK68849.1 |
More Information
Set Membership: |
|

Genomic Map