Assay Details
Target Gene Details
Entrez Gene ID: | 50914 |
Gene Name: | oligodendrocyte transcription factor 1 |
Gene Aliases: |
AI836478, AW494459, Bhlhb6, Olg-1, Oligo1, bHLHe21 |
Location: |
Chr.16:91269769-91271939 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olig1 | NM_016968.4 | 1 | 1054 | NP_058664.2 |
| AB038696.1 | 1 | 1035 | BAB18906.1 | |
| AK144435.1 | 1 | 1052 | BAE25888.1 | |
| AK158791.1 | 1 | 748 | BAE34667.1 | |
| BC046316.1 | 1 | 1013 | AAH46316.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map