Assay Details
Target Gene Details
Entrez Gene ID: | 16443 |
Gene Name: | intersectin 1 (SH3 domain protein 1A) |
Gene Aliases: |
AA517634, AA545208, AI316805, AI839402, AI848451, Ehsh1, Ese1, Itsn, Sh3p17 |
Location: |
Chr.16:91729354-91920591 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itsn1 | NM_001110275.1 | NP_001103745.1 | ||
| NM_001110276.1 | NP_001103746.1 | |||
| NM_010587.2 | NP_034717.2 | |||
| AF132478.1 | AAD19746.1 | |||
| AF132481.1 | AAD19749.1 | |||
| AF468654.1 | AAN32882.1 | |||
| AK155397.1 | BAE33241.1 | |||
| AK160949.1 | BAE36109.1 | |||
| BC066105.1 | AAH66105.1 |
More Information
Set Membership: |
|

Genomic Map