Assay Details
Target Gene Details
Entrez Gene ID: | 16443 |
Gene Name: | intersectin 1 (SH3 domain protein 1A) |
Gene Aliases: |
AA517634, AA545208, AI316805, AI839402, AI848451, Ehsh1, Ese1, Itsn, Sh3p17 |
Location: |
Chr.16:91729354-91920591 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itsn1 | NM_001110275.1 | 20 | 2729 | NP_001103745.1 |
| NM_010587.2 | 20 | 2729 | NP_034717.2 | |
| AF132478.1 | 19 | 2468 | AAD19746.1 | |
| AF132481.1 | 19 | 2468 | AAD19749.1 | |
| AK160794.1 | 5 | 718 | BAE36017.1 | |
| AY127576.1 | 4 | 362 | AAN02285.1 | |
| AY605093.1 | 1 | 185 | AAT36647.1 | |
| AY605094.1 | 4 | 355 | AAT36648.1 |
More Information
Set Membership: |
|

Genomic Map