Assay Details
Target Gene Details
Entrez Gene ID: | 16443 |
Gene Name: | intersectin 1 (SH3 domain protein 1A) |
Gene Aliases: |
AA517634, AA545208, AI316805, AI839402, AI848451, Ehsh1, Ese1, Itsn, Sh3p17 |
Location: |
Chr.16:91729354-91920591 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itsn1 | NM_001110275.1 | 22 | 3101 | NP_001103745.1 |
| NM_010587.2 | 22 | 3101 | NP_034717.2 | |
| AF132478.1 | 21 | 2840 | AAD19746.1 | |
| AF132481.1 | 21 | 2840 | AAD19749.1 | |
| AF169621.1 | AAD48848.1 | |||
| AF498310.1 | AAN32913.1 | |||
| AF498311.1 | AAN32914.1 | |||
| AK082606.1 | BAC38546.1 | |||
| AK160794.1 | 7 | 1090 | BAE36017.1 |
More Information
Set Membership: |
|

Genomic Map