Assay Details
Target Gene Details
Entrez Gene ID: | 56176 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class P |
Gene Aliases: |
Dcrc, Dscr5 |
Location: |
Chr.16:94358763-94371015 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigp | NM_001159616.1 | NP_001153088.1 | ||
| NM_001159617.1 | NP_001153089.1 | |||
| NM_001159618.1 | NP_001153090.1 | |||
| NM_001159619.1 | NP_001153091.1 | |||
| NM_001159620.1 | NP_001153092.1 | |||
| NM_019543.3 | NP_062416.1 | |||
| AF216306.1 | AAF32294.1 | |||
| AF216307.1 | AAF32295.1 | |||
| AF216308.1 | AAF32296.1 | |||
| AF216309.1 | AAF32297.1 | |||
| AK004169.1 | BAB23204.1 | |||
| BC038252.1 | AAH38252.1 | |||
| BC061116.1 | AAH61116.1 | |||
| BC061117.1 | AAH61117.1 | |||
| BC096569.1 | AAH96569.1 | |||
| BX511452.1 | ||||
| BY041400.1 | ||||
| CB596599.1 |
More Information
Set Membership: |
|

Genomic Map