Assay Details
Target Gene Details
Entrez Gene ID: | 14450 |
Gene Name: | phosphoribosylglycinamide formyltransferase |
Gene Aliases: |
Gaps, Prgs |
Location: |
Chr.16:91621395-91647481 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gart | NM_010256.2 | NP_034386.2 | ||
| AK051812.1 | 1 | 2997 | ||
| AK146355.1 | BAE27107.1 | |||
| AK146478.1 | ||||
| AK168445.1 | BAE40349.1 | |||
| AK168458.1 | ||||
| AK168501.1 | BAE40386.1 | |||
| AK168724.1 | BAE40565.1 | |||
| AK168796.1 | BAE40629.1 | |||
| AK168864.1 | BAE40683.1 | |||
| AK168876.1 | BAE40694.1 | |||
| AK177198.1 | ||||
| AK183944.1 | ||||
| AK204723.1 | ||||
| AK205057.1 | ||||
| BB843046.1 | ||||
| BC070465.1 | AAH70465.1 | |||
| U01023.1 | AAA19012.1 | |||
| U01024.1 | AAA19013.1 |

Genomic Map