Assay Details
Target Gene Details
Entrez Gene ID: | 110749 |
Gene Name: | chromatin assembly factor 1, subunit B (p60) |
Gene Aliases: |
2600017H24Rik, C76145, CAF-I p60, CAF-Ip60, CAF1, CAF1A, CAF1P60, MPHOSPH7 |
Location: |
Chr.16:93883895-93906115 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chaf1b | NM_028083.4 | NP_082359.1 | ||
| AK011243.1 | BAB27490.1 |
More Information
Set Membership: |
|

Genomic Map