Assay Details
Target Gene Details
Entrez Gene ID: | 224440 |
Gene Name: | SET domain containing 4 |
Gene Aliases: |
AY037804, C21orf18, ORF21 |
Location: |
Chr.16:93583457-93604833 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Setd4 | NM_145482.3 | 12 | 1688 | NP_663457.2 |
| AK137485.1 | 3 | 2481 | ||
| AY037804.1 | 12 | 1688 | AAK68849.1 | |
| BY701902.1 | 3 | 227 |
More Information
Set Membership: |
|

Genomic Map